Imprimer
Kits for Precellys®
Fournisseur: Bertin Instruments
P000925-LYSK0A P000926-LYSK0A P000927-LYSK0A P000928-LYSK0A P000929-LYSK0A P000930-LYSK0A P000931-LYSK0A P000940-LYSK0A P000910-LYSK0A P000911-LYSK0A P000912-LYSK0A P000914-LYSK0A P000916-LYSK0A P000917-LYSK0A P000918-LYSK0A P000919-LYSK0A P000922-LYSK0A P000923-LYSK0A P000924-LYSK0A P000936-LYSK0A P000937-LYSK0A P000938-LYSK0A P000943-LYSK0A P000944-LYSK0A P000946-LYSK0A P000951-LYSK0A P000952-LYSK0A P000932-LYSK0A P000933-LYSK0A P000934-LYSK0A P000947-LYSK0A P000961-LYSK0A P000948-LYSK0A P000949-LYSK0A P000939-LYSK0A P000935-LYSK0A P000913-LYSK0A P000945-LYSK0A P000950-LYSK0A P000973-LYSK0A P000972-LYSK0A P002137-LYSK0A D34200.10 P002136-LYSK0A
432-0355EA 300 EUR
432-0355 432-0357 432-0356 432-0295 432-0296 432-0297 432-0298 432-0289 432-3753 432-3752 432-3751 432-3754 432-0141 432-0142 431-0170 432-0285 432-0286 432-0287 432-0305 432-0143 432-0144 432-0145 432-0352 432-0353 432-0354 432-0306 432-0421 432-0362 432-0293 432-0294 432-0276 432-0350 432-0277 432-0309 432-0288 432-0292 432-3755 432-0351 432-0278 432-0364 432-0363 710-2071 432-0445 710-2070
Kits for Precellys®
Homogénéisateurs Homogénéisateurs de paillasse
The Precellys lysing kits offer the best versatility of volumes and grinding material to process any type of samples: Human, animal, plants, microorganisms or soil.
  • More than 30 different lysing matrixes
  • Reinforced tubes are suitable for use at the maximum energy levels
  • Easy-handling features allow the Precellys to be used without any advanced training
These kits are specifically for sample preparation for Precellys homogenisers. They can process any type of sample. Choosing the appropriate lysing kit maximises the quantity and quality of extractionMore Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR