Imprimer
Culture tubes, disposable, KIMBLE®
Fournisseur: DWK Life Sciences
SP73500-25150 SP73500-18150 SP73500-16100A
392-0368EA 193 EUR
392-0368 392-0367 SCERSP73500-16100A
Culture tubes, disposable, KIMBLE®
Tubes Tubes de culture
Culture tubes made from 51 expansion borosilicate glass that offer excellent chemical resistance against sodium leaching, a factor common in soda lime glass products.
  • Features sturdy, uniform bottoms and consistent lengths
  • Suitable for cell washing procedures
  • Plain and disposable
Certifications: Glass meets ASTM E438, Type I, Class B requirements. Designed from ASTM E890 specifications.
Conditionnement: Tubes are packed in convenient trays that are shrink-wrapped in plastic film, with tubes remaining in constant alignment for protection against in-More Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR