Imprimer
Cooled Incubators, KB Series
Fournisseur: Binder
9020-0471 9020-0472 9020-0473 9020-0474 9020-0475 9020-0476
390-1741EA 7400 EUR
390-1741 390-1742 390-1743 390-1744 390-1745 390-1746
Cooled Incubators, KB Series
Incubateurs Incubateurs réfrigérés
The powerful KB Series of cooling incubators for microorganisms are able to incubate with exceptional precision even when fully loaded. Units have a stainless-steel inner chamber, feature proven APT.line™ preheating chamber technology, have a temperature range of 0 to +70 °C, hot-air disinfection routine, and can be programmed for weekend operation with ease. This KB unit consumes as much as 40% less energy than the previous model.
  • Energy-efficient cooling with modern inverter cooling unit
  • Cooling with climate-neutral R-600a refrigerant (GWP is 3)
  • Door
More Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR