Imprimer

Bromide, Ion chromatography standard solution, Specpure®, Br- 1000µg/ml

Fournisseur: Thermo Fisher Scientific

35552.AE 35552.AP
35552.AEEA 157 EUR
35552.AE 35552.AP
Bromide, Ion chromatography standard solution, Specpure®, Br- 1000µg/ml
Étalons Étalons anioniques Standards de bromure

Molecular Formula: KBr in H₂O
Physical Form: Liquid
MDL No.: MFCD00134172

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR