Imprimer

Baryum chlorure dihydraté 99, Puriss. p.a. plus Reag. Ph. Eur., ACS, ISO analytical reagent, Fluka™

Fournisseur: Honeywell Chemicals
Danger

Synonyms: Barium dichloride

31125-100G 31125-1KG 31125-2.5KG 31125-4X5KG 31125-500G 31125-5KG 31125-6X1KG
31125-100GEA 74.8 EUR
31125-100G 31125-1KG 31125-2.5KG 31125-4X5KG 31125-500G 31125-5KG 31125-6X1KG
Baryum chlorure dihydraté 99, Puriss. p.a. plus Reag. Ph. Eur., ACS, ISO analytical reagent, Fluka™
Baryum chlorure dihydraté

  • Exceeds standard ACS specifications with additional trace metals analysis
  • Products with a guarantee certificate and/or suitable for the stated analytical application.

Formule: BaCl₂·2H₂O
MW: 244.27 g/mol
Point d'ébullition: 1.560 °C (1.013 hPa)
Point de fusion: 962 °C
Densité: 3.1 g/cm³ (20 °C)
Température de stockage: Température ambiante
Numéro MDL: MFCD00149154
Numéro CAS: 10326-27-9
EINECS: 233-788-1
ONU: 1564
ADR: 6.1 III
Merck Index: 14,00971

Commander maintenant

Résultats des tests de spécification

Assay Min. 99.0 %
Loss on drying (150°C) 14.0 - 16.0 %
Water insoluble matter Max. 0.005 %
pH (5 %, 25 °C) 5.2 - 8.0
Calcium (Ca) Max. 0.005 %
Iron (Fe) Max. 1 ppm
Potassium (K) Max. 0.0025 %
Magnesium (Mg) Max. 0.001 %
Sodium (Na) Max. 0.005 %
Lead (Pb) Max. 0.001 %
Strontium (Sr) Max. 0.01 %
Heavy metals (ICP) ACS Max. 5 ppm
Oxidizing substances (as NO₃) Max. 0.005 %
Total N Max. 0.002 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR