Imprimer

Supelco® PESTANAL® Organic Reference Standard, Tiadinil

Fournisseur: Merck

30698-100MG
30698-100MGEA 295 EUR
30698-100MG
Supelco® PESTANAL® Organic Reference Standard, Tiadinil
Étalons Étalons organiques

  • GHS signal: GHS07


Conditionnement: Bottomless glass bottle. Contents are inside inserted fused cone.

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8eeabe1b7322f423f1606a36186490976b6d9a14&vid=8eeabe1b7322f423f1606a36186490976b6d9a14&prid=web&clid=SAPDAM

Commander maintenant

SPÉCIFICATIONS

Synonyme(s) 5-(3-Chloro-4-methylanilinocarbonyl)-4-methyl-1,2,3-thiadiazole; N-(3-Chloro-4-methylphenyl)-4-methyl-1,2,3-thiadiazole-5-carboxamide
Environmentally Preferable
M-Clarity quality level MQ100
N ° CAS. 223580-51-6
Formula C₁₁H₁₀ClN₃OS
Molecular weight 267,73
Point de fusion 111...113 °C
Description de la classe de danger H332
Point d'éclair Not applicable
Conditions de transport STD
Conditions de stockage 2...30 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR