Imprimer
ExoBrite™ EV Surface Stain Sampler Kit
Fournisseur: Biotium
30127
30127.EA 517 EUR
30127.
ExoBrite™ EV Surface Stain Sampler Kit
Systèmes de liaison et de marquage des protéines
The ExoBrite™ EV Surface Stain Sampler Kit, Green was developed to offer each of Biotium’s ExoBrite™ EV Surface Stains (CTB, WGA, and Annexin V) for assessing which stain offers the best coverage for the EV samples of interest.
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR