Imprimer
Polyurethane surface sampling bags with PolySponge™, Whirl-Pak®
Fournisseur: NASCO FORT ATKINSON
B01590WA
300-0672EA 543 EUR
300-0672
Polyurethane surface sampling bags with PolySponge™, Whirl-Pak®
Sachets Sachets d'échantillonnage
Make environmental surface sampling faster and easier with Whirl-Pak polyurethane surface sampling bags with premoistened sponge. The durable high-density polyurethane sponge is ready to use and resists tearing and fraying.
  • Each sponge is packed in a sterile, sealed write-on bag
  • Write-on label makes identifying samples easy and convenient
  • Sponge is hydrated with 10 ml of HiCap™ neutralizing broth
  • Terminally gamma processed to guarantee sterility (+1 Sterilization)
  • Opening Tyvek® Pull Tabs are perpendicular to the opening of the bag: Keeps your hands away
More Product Information
Plus About VWR