Imprimer

Sodium peroxodisulfate 97, purified

Fournisseur: VWR Chemicals
Danger

Synonyms: Sodium peroxidisulphate, Sodium persulphate, Disodium peroxodisulphate, Peroxydisulphuric acid disodium salt

27997.368EA 192 EUR
27997.368
Sodium peroxodisulfate 97, purified
Sodium peroxodisulfate
Formule: Na₂S₂O₈
MW: 238.11 g/mol
Point de fusion: 100
Densité: 2.4 g/cm³ (20 °C)
Température de stockage: Température ambiante
Numéro MDL: MFCD00003501
Numéro CAS: 7775-27-1
EINECS: 231-892-1
ONU: 1505
ADR: 5.1 III
ATTEINDRE: 01-2119495975-15
Merck Index: 13,08729

Commander maintenant

Résultats des tests de spécification

Assay Min. 97 %
Heavy metals (as Pb) Max. 20 ppm
Fe (Iron) Max. 50 ppm
Fixed origin of raw material Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR