Imprimer

Cyclohexane ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS pour analyses

Fournisseur: VWR Collection
Danger

Synonyms: Hexahydrobenzene, Hexamethylene

23224.550-200
XXXX23224.550-200EA 0 EUR
XXXX23224.550-200 23224.360
Cyclohexane ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS pour analyses
Cyclohexane
Formule: C₆H₁₂
MW: 84.16 g/mol
Point d'ébullition: 81 °C (1013 hPa)
Point de fusion: 7 °C
Densité: 0.7785 g/cm³ (20 °C)
Point d'éclair: –18 °C
Température de stockage: Température ambiante
Numéro MDL: MFCD00003814
Numéro CAS: 110-82-7
EINECS: 203-806-2
ONU: 1145
ADR: 3 II
ATTEINDRE: 01-2119463273-41
Merck Index: 13,02752
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a1e51ba998bef126804c3fd25b361be72cd9545&vid=d1e7f75d9f2a944eb15a7740c5254588cc9a4e67&prid=web&clid=SAPDAM

Commander maintenant

Résultats des tests de spécification

Assay (on anhydrous substance) Min. 99.5 %
IR Spectrum Passes test
Readily carbonisable substances Passes test
Boiling point 79 - 81 °C
Colouration Max. 10 APHA
Density (20/4) 0.778 - 0.779
Density (20/20) 0.779 - 0.780
Free acidity Max. 0.0003 meq/g
Solidification point 5.5 - 6.5 °C
Substances coloured by H2SO4 Max. 60 APHA
Benzene Max. 100 ppm
Cyclohexene Max. 100 ppm
Evaporation residue Max. 10 ppm
Water Max. 100 ppm
Al (Aluminium) Max. 0.1 ppm
Ba (Barium) Max. 0.02 ppm
Ca (Calcium) Max. 0.5 ppm
Cd (Cadmium) Max. 0.05 ppm
Co (Cobalt) Max. 0.02 ppm
Cr (Chromium) Max. 0.02 ppm
Fe (Iron) Max. 0.1 ppm
K (Potassium) Max. 0.1 ppm
Mg (Magnesium) Max. 0.05 ppm
Mn (Manganese) Max. 0.02 ppm
Na (Sodium) Max. 1 ppm
Ni (Nickel) Max. 0.02 ppm
Pb (Lead) Max. 0.05 ppm
Sn (Tin) Max. 0.1 ppm
Sr (Strontium) Max. 0.02 ppm
Zn (Zinc) Max. 0.1 ppm
Conforms to ACS Passes test
Conforms to Reag. Ph.Eur. Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR