Imprimer
Stopcocks, needle-valve, straight
Fournisseur: Lenz Laborglas GmbH & CO.KG
02270002
227-3310EA 37.2 EUR
227-3310
Stopcocks, needle-valve, straight
Robinets d'arrêt
Needle-valve stopcocks are made of DURAN® borosilicate glass, with PTFE spindle.
  • Screw cap and retention device made of PP
  • Temperature resistance from −20 to +150 °C
  • Excellent chemical resistance
These stopcocks are ideal for exact dispensing and therefore ideal for glass equipment.
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR