Imprimer
Streptomyces sp. N174 chitosanase (from Streptomyces sp. N174)
Fournisseur: Merck Millipore Calbiochem
220477-10
220477-10EA 418 EUR
220477-10
Streptomyces sp. N174 chitosanase (from Streptomyces sp. N174)
Enzymes
Chitosanase from Streptomyces species is a new class of enzymes with antifungal properties that efficiently catalyses the hydrolysis of acetylated chitosan.
  • Does not hydrolyse chitin or carboxylmethylcellulose
  • Activity increases with decreasing acetylation of chitosan
It acts by splitting the β1,4-glycosidic linkages of GlcNAc-GlcN and GlcNAc-GlcN. Suitable for the efficient preparation of protoplasts of Mucor and other Zygomycetes, allowing the genetic manipulation of these fungi.
Unit definition: One unit is defined as the amount of enzyme that will release 1,0 µmol reducing sugars or glucosamine from chitosan per min at 37 °C, pH 5,5.
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR