Imprimer
Sample dippers, DispoDipper
Fournisseur: BURKLE
5393-7000 5393-7001 5393-7010 5393-7011 5393-7020 5393-7021
331-0313EA 42.7 EUR
331-0313 331-0314 216-2206 216-2207 216-2208 216-2209
Sample dippers, DispoDipper
Échantillonneurs Préleveurs d'échantillons
The DispoDipper are designed for the sampling and subsequent transport of samples.
  • Production and packaging according to cleanroom class 7
  • Prevents cross-contamination or sample impurities
  • Optionally sterilised by gamma rays (SteriPlast®)
Suitable for applications in food, animal feed and pharmaceutical industries.
Certifications: EU food requirements and FDA regulations.
Attention: *Le(s) produit(s) sur cette page sont hors du champ d'application de la directive européenne 2019/904
Plus About VWR