Imprimer
Vydac 214ATP C4, HPLC columns
Fournisseur: Avantor
214ATP5405EA 822 EUR
214ATP5405 214ATP54 214ATP5415 214AGD54/N
Vydac 214ATP C4, HPLC columns
Colonnes pour chromatographie
These Vydac TP was the first range of wide pore media phases developed in the Vydac range and became the industry standard for peptide and protein separations. The large pores of the 300 Å TP silica give polypeptide molecules complete access to the interior of the silica pores.
  • Long column lifetime and negligible phase leaching
  • Reliable protein purifications, scalable from analytical to preparative scale
  • Referenced in a large number of patents and publications
214ATP C4 has a lower level of endcapping and is optimised for the analysis of human growthMore Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR