Imprimer
Test tubes, round bottom
Fournisseur: DELTALAB
916150 818150 820150 801075 801275 901275 913100 816150 916100
212-3308EA 123 EUR
212-3308 212-3310 212-3312 212-3315 212-3317 DELT901275 DELT913100 DELT816150 DELT916100
Test tubes, round bottom
Tubes Tubes à essais
Tubes are made of borosilicate or soda glass.
  • Good chemical resistance
  • Good temperature resistance
High quality of tubes is reflected in uniformity of wall thickness, diameter and height dimensions.
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR