Imprimer

Acide fluorhydrique 40%, AnalaR NORMAPUR® zA

Fournisseur: VWR Chemicals
Danger

20307.290EA 48.3 EUR
20307.290 20307.320
Acide fluorhydrique 40%, AnalaR NORMAPUR® zA
Acide fluorhydrique
Formule: HF
MW: 20.01 g/mol
Point d'ébullition: 112 °C (1013 hPa)
Point de fusion: –44 °C
Température de stockage: Température ambiante
Numéro MDL: MFCD00011346
Numéro CAS: 7664-39-3
EINECS: 231-634-8
ONU: 1790
ADR: 8 (6.1) II

Commander maintenant

Résultats des tests de spécification

Assay 39.0 - 41.0 %
Heavy metals (as Pb) Max. 0.5 ppm
SiF6 (Hexafluorosilicate) Max. 50 ppm
Cl (Chloride) Max. 5 ppm
PO4 (Phosphate) Max. 1 ppm
SO4 + SO3 (as SO4) Max. 5 ppm
Ag (Silver) Max. 0.02 ppm
Al (Aluminium) Max. 0.05 ppm
As (Arsenic) Max. 0.05 ppm
Ba (Barium) Max. 0.1 ppm
Be (Beryllium) Max. 0.02 ppm
Bi (Bismuth) Max. 0.1 ppm
Ca (Calcium) Max. 0.5 ppm
Cd (Cadmium) Max. 0.01 ppm
Co (Cobalt) Max. 0.02 ppm
Cr (Chromium) Max. 0.02 ppm
Cu (Copper) Max. 0.02 ppm
Fe (Iron) Max. 0.2 ppm
Ge (Germanium) Max. 0.05 ppm
Li (Lithium) Max. 0.02 ppm
Mg (Magnesium) Max. 0.2 ppm
Mn (Manganese) Max. 0.05 ppm
Mo (Molybdenum) Max. 0.05 ppm
K (Potassium) Max. 0.1 ppm
Na (Sodium) Max. 0.2 ppm
Ni (Nickel) Max. 0.02 ppm
Pb (Lead) Max. 0.05 ppm
Sr (Strontium) Max. 0.02 ppm
Ti (Titanium) Max. 0.1 ppm
Tl (Thallium) Max. 0.05 ppm
V (Vanadium) Max. 0.05 ppm
Zn (Zinc) Max. 0.05 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR