Imprimer
Diaphragm Vacuum Pumps and Compressors
Fournisseur: KNF
N022AN.18 N022AT.18/0003
181-3212EA 669 EUR
181-3212 KNFAN022AT.18/0003
Diaphragm Vacuum Pumps and Compressors
Pompe Pompes à vide à membrane
Series N 022 diaphragm pumps are single-head, dry-running devices used in a wide range of laboratory applications. They transfer, compress and pump down without contamination.
  • 100% oil-free transfer
  • Pure transferring, evacuation and compression of gases
  • Version for slightly aggressive or corrosive gases and vapours
  • Maintenance-free
  • Gastight, leakage rate approx. 6×10⁻³ mbar×L/s, not tested in serial production
The pumps are available in various versions differing in the materials which contact the media.
Connectors for tube: 6 int.Ø
Motor protection:More Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR