Imprimer
Milli-Q® IQ Element purification system
Fournisseur: Merck
IPAKICPK1 ZIQELEMT0
172-0054EA 1720 EUR
172-0054 172-0063
Milli-Q® IQ Element purification system
Purificateurs d'eau Systèmes d'eau pure et ultra-pure
Milli-Q® IQ Element water purification and dispensing unit is designed to fit into your trace analysis workflow. The Milli-Q® IQ Element unit, combined with a Milli-Q® IQ 7 series water purification system, delivers analytical grade ultrapure water that is suitable for trace and ultra-trace elemental analyses, including ICP-MS, GF-AAS and trace IC.
  • Designed to fit into your trace analysis workflow
  • Easy to integrate – the compact unit is designed for seamless, contaminant-free installation in your cleanroom environment or laminar flow hood
  • Easy to use – a
More Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR