Imprimer
Organic reference standard, Bisphenol F bis(3-chloro-2-hydroxypropyl) ether, mixture of 3 isomers ortho-ortho, ortho-para, para-para, Supelco®
Fournisseur: Merck
15139-250MG
15139-250MGEA 207 EUR
15139-250MG
Organic reference standard, Bisphenol F bis(3-chloro-2-hydroxypropyl) ether, mixture of 3 isomers ortho-ortho, ortho-para, para-para, Supelco®
Étalons Étalons organiques
Bisphenol F bis(3-chloro-2-hydroxypropyl) ether a additive used during synthesis of organosol resin preventing theremal degradation of polymer. During the removal of hydrochloric acid in organosol resin results in this chlorinated compound.
  • Used as reference standard in RP-HPLC-APCI-MS-MS analysis for optimising and quantifying pressurised liquid extraction for BFDGE and its derivatives from canned food
Standard for determining toxic monomers released from polymers of the inner coating of cans.
Conditionnement: Bottomless glass bottle. Contents are inside inserted fused cone.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=52538f24a14c469801d093ed2fbff37f841b959f&vid=52538f24a14c469801d093ed2fbff37f841b959f&prid=web&clid=SAPDAM
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR