Imprimer

Magnesium standard dissolved in oil, 1,000 µg/g Mg in mineral oil Specpure® for wear metal analysis

Fournisseur: Thermo Fisher Scientific

14219.1Y 14219.18
14219.1YEA 0 EUR
14219.1Y 14219.18
Magnesium standard dissolved in oil, 1,000 µg/g Mg in mineral oil Specpure® for wear metal analysis
Étalons Étalons cationiques Standards de magnésium

Physical Form: Liquid
MDL No.: MFCD00085308

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR