Imprimer

Strontium standard dissolved in oil, 1,000 µg/g Sr in mineral oil Specpure® for wear metal analysis

Fournisseur: Thermo Fisher Scientific

14205.18
14205.18EA 91.7 EUR
14205.18
Strontium standard dissolved in oil, 1,000 µg/g Sr in mineral oil Specpure® for wear metal analysis
Étalons Étalons cationiques Standards de strontium

Physical Form: Liquid
MDL No.: MFCD00134060

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR