Imprimer
Safety cabinets, type 90, with wing door technology, CLASSIC line standard and CLASSIC line pro
Fournisseur: DUPERTHAL
29-200667-030 29-200967-030 29-201267-030 29-201267-032 29-200667-030L 29-200667-430L 29-130667-430 29-131267-030 29-130667-030 29-130667-030L 29-201267-050 29-201267-051 29-201267-052 29-201267-053 29-201267-056 29-201267-057 29-201267-058 29-201267-059 29-200967-050 29-200967-051 29-200967-052 29-200967-053 29-200677-053 29-200667-050 29-200667-051 29-200667-052 29-200667-053 29-200677-053L 29-200667-050L 29-200667-051L 29-200667-052L 29-200667-053L 29-201267-430 29-201267-432 29-200967-430 29-200667-430 29-130667-430L 29-131267-430 29-201267-450 29-201267-451 29-201267-452 29-201267-453 29-201267-459 29-200967-450 29-200967-451 29-200967-452 29-200967-453 29-200667-451 29-200667-452 29-200667-453 29-200677-453L 29-200667-450L 29-200667-451L 29-200667-452L 29-200667-453L 29-201265-030
135-1958EA 2239.92 EUR
135-1958 135-1685 132-1308 132-1309 135-2501 135-2511 132-1304 132-1306 132-1302 132-1303 132-1310 132-1312 132-1313 132-1314 132-1315 132-1316 132-1317 132-1318 135-2524 135-2525 135-2526 135-2527 135-2520 135-2502 135-2504 135-2506 135-2508 135-2521 135-2503 135-2505 135-2507 135-2509 132-1319 132-1320 135-2528 135-2510 132-1305 132-1307 132-1321 132-1322 132-1323 132-1324 132-1328 135-2529 135-2530 135-2531 135-2532 135-2514 135-2516 135-2518 135-2523 135-2513 135-2515 135-2517 135-2519 DUEH29-201265-030
Safety cabinets, type 90, with wing door technology, CLASSIC line standard and CLASSIC line pro
Mobilier Armoires de stockage
Safety cabinets with storage shelves (CLASSIC line standard) or pull-out shelves (CLASSIC line pro) for the storage of hazardous substances in working spaces. Complete carcass, interior fittings and wing doors are made of powder-coated sheet steel.
  • Doors with standard integrated arrest system, meaning that containers can be placed into and removed from the cabinet with both hands
  • Removable base and adjustable feet for easy alignment and transport with a hand pallet truck.
  • The base is set back to prevent hitting one’s feet
  • Ventilation at every cabinet
More Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR