Imprimer
PureClean™ Tyvek® 1421B Self-Sealing Autoclave Bags with Steam Indicator
Fournisseur: KEYSTONE CLEANROOM PRODUCTS
6STKRP0428SS 6STKRP0430SS 6STKRP0608SS 6STKRP0610SS 6STKRP0612SS 6STKRP0615SS 6STKRP0616SS 6STKRP0638SS 6STKRP0707SS 6STKRP1216SS 6STKRP1218SS 6STKRP1220SS 6STKRP1222SS 6STKRP1224SS 6STKRP1228SS 6STKRP1436SS 6STKRP1438SS 6STKRP1440SS 6STKRP1448SS 6STKRP1526SS 6STKRP1543SS 6STKRP1604SS 6STKRP1612SS 6STKRP1616SS 6STKRP1240SS 6STKRP1242SS 6STKRP1246SS 6STKRP1248SS 6STKRP1414SS 6STKRP1416SS 6STKRP1418SS 6STKRP1824SS 6STKRP0713SS 6STKRP0811SS 6STKRP0815SS 6STKRP0816SS 6STKRP0820SS 6STKRP0838SS 6STKRP0843SS 6STKRP0709SS 6STKRP09-512SS 6STKRP1015SS 6STKRP1018SS 6STKRP1020SS 6STKRP1022SS 6STKRP1024SS 6STKRP1030SS 6STKRP1036SS 6STKRP1048SS 6STKRP1056SS 6STKRP1062SS 6STKRP1212SS 6STKRP1214SS 6STKRP1230SS 6STKRP1232SS 6STKRP1234SS 6STKRP1236SS 6STKRP3456SS 6STKRP3624SS 6STKRP3628SS 6STKRP3636SS 6STKRP3642SS 6STKRP3646SS 6STKRP1420SS 6STKRP1424SS 6STKRP1426SS 6STKRP1620SS 6STKRP1624SS 6STKRP1628SS 6STKRP1630SS 6STKRP1632SS 6STKRP1635SS 6STKRP1636SS 6STKRP1638SS 6STKRP1644SS 6STKRP1648SS 6STKRP1820SS 6STKRP2024SS 6STKRP2026SS 6STKRP2030SS 6STKRP2032SS 6STKRP2036SS 6STKRP2040SS 6STKRP2044SS 6STKRP2046SS 6STKRP2048SS 6STKRP1828SS 6STKRP1830SS 6STKRP1832SS 6STKRP1836SS 6STKRP1840SS 6STKRP1848SS 6STKRP1854SS 6STKRP1870SS 6STKRP2020SS 6STKRP2134SS 6STKRP2462SS 6STKRP2628SS 6STKRP2634SS 6STKRP2646SS 6STKRP2828SS 6STKRP2832SS 6STKRP2834SS 6STKRP2840SS 6STKRP2844SS 6STKRP2224SS 6STKRP2226SS 6STKRP2240SS 6STKRP2260SS 6STKRP2424SS 6STKRP2430SS 6STKRP2436SS 6STKRP2440SS 6STKRP2448SS 6STKRP2452SS 6STKRP3016SS 6STKRP3020SS 6STKRP3028SS 6STKRP3030SS 6STKRP3036SS 6STKRP3060SS 6STKRP3232SS 6STKRP3238SS 6STKRP3248SS 6STKRP3412SS 6STKRP3434SS 6STKRP3838SS 6STKRP4024SS 6STKRP4040SS 6STKRP4048SS 6STKRP4054SS 6STKRP4060SS 6STKRP4072SS 6STKRP4242SS 6STKRP4244SS 6STKRP3648SS 6STKRP3660SS 6STKRP3664SS 6STKRP3672SS 6STKRP4416SS 6STKRP4450SS 6STKRP4848SS 6STKRP4850SS 6STKRP4864SS 6STKRP5054SS 6STKRP6250SS 6STKRP0426SS 6STKRP0410SS 6STKRP0406SS 6STKRP0409SS 6STKRP0404SS 6STKRP0416SS 6STKRP0205SS 6STK0308TD 6STK2224SS
129-4091EA 2681 EUR
129-4091 129-4159 129-4145 129-4182 129-4168 129-4046 129-4174 129-4143 129-4103 129-4055 129-4074 129-4063 129-4189 129-4113 129-4058 129-4185 129-4104 129-4075 129-4151 129-4150 129-4160 129-4054 129-4129 129-4050 129-4087 129-4183 129-4111 129-4053 129-4102 129-4157 129-4083 129-4123 129-4152 129-4181 129-4172 129-4066 129-4195 129-4134 129-4110 129-4155 129-4156 129-4062 129-4132 129-4162 129-4198 129-4052 129-4098 129-4070 129-4122 129-4112 129-4077 129-4100 129-4089 129-4124 129-4148 129-4177 129-4088 129-4096 129-4140 129-4161 129-4199 129-4120 129-4171 129-4193 129-4080 129-4164 129-4117 129-4064 129-4141 129-4158 129-4076 129-4179 129-4187 129-4108 129-4133 129-4085 129-4114 129-4136 129-4180 129-4186 129-4079 129-4127 129-4192 129-4176 129-4105 129-4093 129-4115 129-4131 129-4142 129-4144 129-4047 129-4068 129-4059 129-4069 129-4048 129-4090 129-4061 129-4065 129-4060 129-4071 129-4169 129-4184 129-4078 129-4128 129-4146 129-4153 129-4056 129-4154 129-4137 129-4119 129-4125 129-4167 129-4067 129-4194 129-4166 129-4099 129-4165 129-4138 129-4094 129-4170 129-4095 129-4139 129-4175 129-4190 129-4173 129-4188 129-4072 129-4057 129-4126 129-4051 129-4196 129-4191 129-4107 129-4106 129-4101 129-4147 129-4116 129-4109 129-4178 129-4097 129-4118 129-4092 129-4049 129-4197 129-4081 129-4073 129-4082 129-4084 129-4086 129-4121 129-4135 129-4149 129-4163 KACC6STK0308TD KACC6STK2224SS
PureClean™ Tyvek® 1421B Self-Sealing Autoclave Bags with Steam Indicator
Sachets Poches de stérilisation
PureClean™ self-sealing autoclave bags are made from a transparent film and 1421B DuPont™ Tyvek®, which is a new grade of uncoated, untreated, flexible Tyvek® material specifically designed for pharmaceutical use in steam sterilisation applications. Uncoated Tyvek® eliminates the potential of residue deposits on equipment, post sterilisation.
  • Clear side enables visual confirmation of dryness for sterilised parts and equipment, which is required by EU GMP Annex 1
  • Convenient self-seal closure eliminates the need for autoclave tape or the validation and use
More Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR