Imprimer

VWR® Autoclave Bags, Polypropylene

Fournisseur: VWR Collection
129-4026EA 150 EUR
129-4026 129-4027 129-4030 129-4031 129-4032 129-4033 129-4034 129-4035 129-4036 129-4038 129-4039 129-4040 129-4042 129-4043
VWR® Autoclave Bags, Polypropylene
Sachets Sacs à déchets

Sturdy, temperature resistant bags, ideal for disposing of non-hazardous waste or hazardous waste that must first be autoclaved. Can also be used for autoclaving other products. Supplied complete with wire closures that allow expansion of the bag and ventilation when cooling.



  • Autoclavable up to 135 °C maximum

  • Include 200 mm twist ties which allow bag to expand and vent steam during cooling

  • Bags meet the requirements of the 165 g puncture inspection standard as described in ASTM D1709
  • Available in different thickness: 37,5, 50  or 76 µm thick
  • Available
More Product Information
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR