Imprimer
PIG® Rip-&-Fit® HazMat rolls
Fournisseur: NEW PIG
MAT342 MAT343
121-0530EA 88.5 EUR
121-0530 NEWPMAT343
PIG® Rip-&-Fit® HazMat rolls
Matériaux absorbants
Extra perforations let you create custom sizes of PIG HazMat Mat without cutting. Just "rip" it along the easy-tear perfs and "fit" it into place.
  • Absorbs the widest range of acids, caustics and unknown liquids - even those with high concentrations like 98% sulfuric acid and 30% sodium hydroxide
  • Chemical-resistant mat won't degrade or cause a dangerous reaction upon contact with corrosive spills
  • Extra perforations in our Rip-&-Fit Mat allow you to easily "rip" this PIG Mat and "fit" it to your application without cutting
  • No matter how you rip it, this
More Product Information
Plus About VWR