Imprimer
PIG® medium and large mobile container kits
Fournisseur: NEW PIG
KITE203 KITE302 KITE303 KITE402 KITE403 RFLE202 RFLE203 RFLE302 RFLE303 RFLE402 RFLE403 KITE202
121-0063EA 687 EUR
121-0063 121-0068 121-0069 121-0072 121-0073 121-0118 121-0119 121-0123 121-0124 121-0126 121-0127 121-0062
PIG® medium and large mobile container kits
Matériaux absorbants
Fast, effective response to indoor and outdoor spills.
  • Durable, wheeled 240 and 360 L container is packed with high-quality PIG® absorbents
  • Easy to transport supplies directly to the spill site
  • Hinged lid makes it easy to get the absorbents you need — just flip it open
  • Container with bold Spill Kit lettering for quick identification
  • Absorbents are organised for quick, easy response
  • Heavy-duty rubber wheels roll easily over nearly any surface
Weather-resistant polyethylene container protects the contents from moisture, dirt and damage (containers areMore Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR