Imprimer
Flash Chromatography Systems, C-8XX PURE
Fournisseur: BUCHI
11C80500 11C81000 11C81500 11C83000 11C83500 11C85000
11C80500.EA 21200 EUR
11C80500. 11C81000. 11C81500. 11C83000. 11C83500. 11C85000.
Flash Chromatography Systems, C-8XX PURE
Systèmes de chromatographie Systèmes de flash-chromatographie
PURE chromatography systems for flash applications.
  • Benefit from convenient placement of the cartridge/column holder
  • Gain extra bench space with solvent platform
  • Enjoy clean air using a fume enclosure
  • Feel safe with your system thanks to automatic monitoring and settings
  • Perform flexible sample injection thanks to smart design
  • Minimise user training time thanks to simple software
Perform any flash or prep HPLC application easily and safely with our extremely compact pure chromatography systems. The pure platform is fitting to your needs andMore Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR