Imprimer
Safety cans, type II AccuFlow™, galvanised steel
Fournisseur: JUSTRITE MANUFACTURING
7210120Z 7225130Z 7220120Z 7250130Z 7220220Z 7250230Z
119-0418EA 119 EUR
119-0418 119-0421 119-0419 119-0424 119-0420 119-0425
Safety cans, type II AccuFlow™, galvanised steel
Bidons de sécurité
Safety cans are designed to convenient and safe storage and dispense of flammable liquids.
  • Rounded steel comfort handle protects hand injury
  • Trilingual warning label in English/Spanish/French
  • Large ID zone identifies contents to reduce misuse
Features vented AccuFlow™ manifold and Safe-Squeeze® trigger handle provides the controlled liquid flow ever and flexible metal hose of 229 mm long to reach small openings. Ergonomically designed quick-fill port with comfort lift lever makes filling easier than ever.
Fill port is equipped with a long length,More Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR