Imprimer
Gas detection tubes for carbon disulphide
Fournisseur: Dräger
8101891 6728351 CH23201
151-0183EA 116 EUR
151-0183 DRAE6728351 118-0339
Gas detection tubes for carbon disulphide
Tubes de détection de gaz
Operating conditions: 0 to 40 °C ; up to 30 mg/l H₂O and for tube 5/a: 3 to 15 mg/l H₂O absolute humidity.
  • Data below representative for use with an Accuro® pump
  • Cross sensitivity for tube 3/a hydrogen sulphide in the TLV range retained in the pre-layer (does not interfere)
  • For tubes carbon disulphide 5/a and 30/a it is impossible to measure carbon disulphide in the presence of hydrogen sulphide
One stroke is equivalent to a volume of 100 ±5 ml of gas passing through the tube.
The standard measuring range is calibrated at 20 °C and 1013 hPa.
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR