Imprimer
Eyewash station, HypaClens
Fournisseur: SAFETY FIRST AID GROUP
E458
118-0283EA 59.9 EUR
118-0283
Eyewash station, HypaClens
Matériel de premiers secours Stations et douches pour le lavage oculaire
This wall-mountable eyewash station holds two 500 ml HypaClens eyewash bottles.
  • Integrated mirror allows for self-administration of eyewash
  • Compact design of the eyewash station makes it ideal for any work place with low risks
W×D×H (mm): 250×60×300
Informations de commande: Station is supplied with two bottles of eyewash solution. Wall fixings are not included. Refills are available separately, please contact your local Avantor sales office for further information.
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR