Imprimer
Lingettes pour salle propre, AlphaWipe®
Fournisseur: Texwipe
TX1004 TX1013 TX1009B TX1009
115-0496EA 120 EUR
115-0496 115-0454 115-1769 115-1949
Lingettes pour salle propre, AlphaWipe®
Lingettes Lingettes pour salles propres Lingettes pour salles propres, sèches
These premium heavy-weight all-purpose wipes are made from 100% continuous filament polyester in a double-knit, no run, interlocked pattern. Soft and non abrasive, they are ideal for critical environments where contamination control is essential.
  • Ultra-low particulate and fibre levels
  • Excellent for sensitive surfaces
  • Superior strength guards against abrasion under rigorous use
Suitable for use in Class 10 to 100000, ISO 4 to 8 cleanrooms.
Couleur: Blanc.
Attention: Data accurate at time of publishing, latest test results and test methods can beMore Product Information
Plus About VWR