Imprimer
VWR® Basic, Frocks, SMS
Fournisseur: VWR Collection
113-1175EA 197 EUR
113-1175 113-1166 113-1174 113-1176 113-1162 113-1161 113-1160 113-1159 113-1158 113-1157 113-1156
VWR® Basic, Frocks, SMS
Vêtements à usage unique
These SMS frocks are manufactured from a latex-free, multi-layer spunbonded - melt-blown - spunbonded (SMS) fabric, that offers high tensile strength while remaining soft, comfortable and breathable.
  • Snap collar, raglan sleeves, snap closure
  • Fluid and particulate barrier
  • Light chemical splash resistance
Colour: White or blue
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR