Imprimer
General purpose gloves, Dexgrip Light
Fournisseur: HONEYWELL SAFETY
2094150/10
112-1469EA 0 EUR
112-1469
General purpose gloves, Dexgrip Light
Gants
These knitted gloves made of cotton and polyamide feature a crinkled latex coating on the palm and fingertips.
  • Good dexterity, maximum comfort and added sturdiness
  • Tight knit (gauge 13) gives the glove perfect support, limits the penetration of dirt and ensures a good touch
  • Cotton thread absorbs perspiration and thus provides greater comfort
  • Ultra fine crinkled latex coating offers maximum dexterity while providing mechanical resistance
  • Good grip, even in a damp environment
Ideal for the handling of slippery, damp or non greasy parts. Suitable forMore Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR