Imprimer
Lunettes-masques, uvex carbonvision
Fournisseur: UVEX
9307.365
111-2535EA 0 EUR
111-2535
Lunettes-masques, uvex carbonvision
Lunettes de protection
Modern materials and manufacturing technologies enable the design of frames which combine reduced size and a high mechanical resistance.
  • TPU layer is permanently fixed to the frame, ensuring a perfect individual fit; easy to clean
  • Lightweight enabling the wearer to work for longer periods without any distractions and problems
  • Flexible soft components adapt perfectly to the face ensuring a perfect fit
  • Convenient clip to remove headband before cleaning
Ces masques sont recommandés comme protection contre la poussière, les éclaboussures de liquides etMore Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR