Imprimer
Reusable half masks, MX/PF
Fournisseur: HONEYWELL SAFETY
1001558
111-0548EA 67.7 EUR
111-0548
Reusable half masks, MX/PF
Protection respiratoire Masques réutilisables Demi-masques
The half mask MX/PF is designed to work when the type of protection depends on the cartridges connected, gases, particles or combined. The APF is evaluated from 12 to 50.
  • Easy assembly
  • Silicone facepiece for additional comfort and easy decontamination
  • Triple flange seal
  • Wide, horizontal skin contact with pleated design
  • Good fit accross a wide variety of facial geometries
The half mask SPERIAN MX PF can be used only with certified T SERIES cartridges (following the standard EN 141 , EN 143)
Recommended for long term use
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR