Imprimer

Zirconium 99,5 Zr + Hf, sponge 0.8-25.4 mm (0.03-1.0 in)

Fournisseur: Thermo Fisher Scientific
Danger

10441.18 10441.30 10441.A1
10441.18EA 74.4 EUR
10441.18 10441.30 10441.A1
Zirconium 99,5 Zr + Hf, sponge 0.8-25.4 mm (0.03-1.0 in)
Zirconium

Purity: 99.5% CAS No.: 7440-67-7 Odour: Odourless UN No.: 2008 MDL No.: MFCD00011303

Formule: Zr
MW: 91.22 g/mol
Température de stockage: Température ambiante
Numéro MDL: MFCD00011303
Numéro CAS: 7440-67-7
EINECS: 231-176-9
ONU: 2008
ADR: 4.2 III

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR