Imprimer
HPLC columns, LiChroCART®, Superspher® 60 RP-8
Fournisseur: Merck
1.50171.7139 1.50173.7139 1.50175.7139 1.50177.7139 1.50178.7139 1.50179.7139 1.50195.7139 1.50201.7139 1.50228.7139 1.50229.7139 1.50233.7139 1.50234.7139 1.50236.7139 1.50302.7139 1.51432.7139 1.51442.7139 1.51443.7139 1.51444.7139 1.51448.7139 1.50170.7139 1.50177.7141
1.50171.7139EA 660 EUR
1.50171.7139 1.50173.7139 1.50175.7139 1.50177.7139 1.50178.7139 1.50179.7139 1.50195.7139 1.50201.7139 1.50228.7139 1.50229.7139 1.50233.7139 1.50234.7139 1.50236.7139 1.50302.7139 1.51432.7139 1.51442.7139 1.51443.7139 1.51444.7139 1.51448.7139 1.50170.7139 1.50177.7141
HPLC columns, LiChroCART®, Superspher® 60 RP-8
Colonnes pour chromatographie
Stainless steel LiChroCART® cartridges are available with various types of Superspher® sorbents.
These LiChroCART® cartridges require part number 1.51486.0001 manu-CART® cartridge column holder, which can be used to hold one cartridge column with or without a  4×4 mm guard column. LiChroCART® columns 250×10 mm require part number 1.51419.0001 manu-CART® 10. The short LiChroCART® columns can be ordered as a set including the corresponding cartridge holder and one cartridge, or as a pack of 3 cartridges without cartridge holder. The separate part numbers for the cartridge are asMore Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR