Imprimer
Silicate CRM, Standard Solutions, Supelco®
Fournisseur: Merck
1.32243.0100 1.32244.0100 1.32245.0100
1.32243.0100EA 55.9 EUR
1.32243.0100 1.32244.0100 1.32245.0100
Silicate CRM, Standard Solutions, Supelco®
Étalons Étalons anioniques Standards de silicate
SiO₂ in H₂O
  • Diluted standard solutions that are ready-to-use for accurate measurements
  • Traceable to SRM from NIST and USP
  • Full certificates of analysis with comprehensive quality documentation
  • Information on concentration and expanded measurement uncertainty
  • Save time and minimize errors
Method used for Uncertainty Measurement (Accuracy in the table below)= Photometry
Certifications: Traceable to SRM from NIST and USP.
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR