Imprimer

Acétonitrile, Prepsolv® Chromatography_prep

Fournisseur: Merck
Danger

Synonyms: Methyl cyanide, Ethyl nitrile, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile

MQ200

1.13358.2500 1.13358.9030 1.13358.9185 1.13358.9910 1.13358.9186 1.13358.9913 1.13358.2509
1.13358.2500EA 243 EUR
1.13358.2500 1.13358.9030 1.13358.9185 1.13358.9910 1.13358.9186 1.13358.9913 1.13358.2509
Acétonitrile, Prepsolv® Chromatography_prep
Acétonitrile

Prepsolv® solvents are tailored to the requirements of preparative HPLC to facilitate scale-up from analytical to preparative separations. With their extremely low evaporation residue (< 1 mg/l) and low water content, they ensure optimal protection for columns.

Formule: H₃CCN
MW: 41.05 g/mol
Point d'ébullition: 81.6 °C (1013 hPa)
Point de fusion: –45.7 °C
Densité: 0.782 g/cm³ (20 °C)
Point d'éclair: 2 °C (coupe fermée)
Température de stockage: Température ambiante
Numéro MDL: MFCD00001878
Numéro CAS: 75-05-8
EINECS: 200-835-2
ONU: 1648
ADR: 3
ATTEINDRE: 01-2119471307-38
Merck Index: 13,00071
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR