Imprimer

Trisodium 2-(4-Sulphophenylazo)-1,8-dihydroxynaphthalene-3,6-disulphonate (SPADNS), GR zA, Supelco®

Fournisseur: Merck

Synonyms: SPADNS, 2-(4-Sulphophenylazo)chromotropic acid trisodium salt, 4,5-Dihydroxy-3-(4-sulphophenylazo)-2,7-naphthalenedisulphonic acidtrisodium salt, 1,8-Dihydroxy-2-(4-sulphophenylazo)naphthalene-3,6-disulphonic acidtrisodium salt, Sulphanilic acid azochromotrope, 2-(p-sulphophenylazo)-1,8-Dihydroxy-3,6-Naphthalenedisulphonic acidtrisodium salt, Trisodium 1,8-Dihydroxy-2-(4-sulphophenylazo)naphthalene-3,6-disulphonate

MQ100

1.07998.0025
1.07998.0025EA 180 EUR
1.07998.0025
Trisodium 2-(4-Sulphophenylazo)-1,8-dihydroxynaphthalene-3,6-disulphonate (SPADNS), GR zA, Supelco®
Trisodium 2-(4-Sulphophenylazo)-1,8-dihydroxynaphthalene-3,6-disulphonate (SPADNS)

Reagent for Fluoride, Zirconium, Thorium

Formule: C₁₆H₁₂N₂O₁₁S₃
MW: 570.40 g/mol
Température de stockage: Température ambiante
Numéro MDL: MFCD00003952
Numéro CAS: 23647-14-5
EINECS: 245-803-9

Commander maintenant

Résultats des tests de spécification

Solubility 220 g/l (22 °C)
Molar Mass 570.4 g/mol
Bulk Density 810 kg/m3
pH 6.0 (10 g/l, H2O, 22 °C)
HS Code 2927 00 00
pH value 6,0
Bulk density 810 kg/m3
Storage temperature +15...+25 °C
Identity passes test
Color change with zirconium passes test pink - violet
Absorption maximum ⋋max 505 - 510 nm
Loss on drying ≤10 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR