Imprimer

Acide citrique, BAKER ANALYZED® ACS, J.T.Baker®

Fournisseur: Avantor
Attention

Synonyms: 2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid

0090.1000EA 116.46 EUR
0090.1000
Acide citrique, BAKER ANALYZED® ACS, J.T.Baker®
Acide citrique
Formule: HOC(COOH)(CH₂COOH)₂
MW: 192.13 g/mol
Point d'ébullition: 310
Point de fusion: 149…151 °C
Densité: 1.665 g/cm³ (20 °C)
Température de stockage: Température ambiante
Numéro MDL: MFCD00011669
Numéro CAS: 77-92-9
EINECS: 201-069-1
ATTEINDRE: 01-2119457026-42-0000
Merck Index: 13,02350
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d60f5a9b8e6c6dfb7eda04e1a1d5d6d971f92444&vid=caf4f9effc7fd22b942f7f92be770b06e40637a4&prid=web&clid=SAPDAM

Commander maintenant

Résultats des tests de spécification

Meets ACS Specifications
Assay min. 99.5 %
Chloride (Cl) max. 0.001 %
Insoluble Matter max. 0.005 %
Oxalate (C2O4) passes test
Phosphate (PO4) max. 0.001 %
Residue after Ignition max. 0.02 %
Sulfur Compounds (as SO4) max. 0.002 %
Substances Carbonizable by Hot H2SO4 (Tartrates, etc.) passes test
Trace Impurities (in ppm)Iron (Fe) max. 3
Trace Impurities (in ppm)Lead (Pb) max. 2


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR